Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCAGAGTGGGGAGCGGCCTTGGTGA[C/T]GGGATTTCTGGGTCCCGTAGACCCT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 114840 MIM: 604890 | ||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
CEL PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese)
|
||||||
EUR
|
||||||||
AMR
|
CEL - carboxyl ester lipase | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001807.4 | Intron | NP_001798.2 |
GTF3C5 - general transcription factor IIIC subunit 5 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
LOC100996574 - uncharacterized LOC100996574 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
HapMap |