Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AATGAGTCTGAGAGACAGGCTTTAT[C/T]TTCTGAAAGGGCCTGTTCAGTTCTG
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 609187 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
SUGCT PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
SUGCT - succinyl-CoA:glutarate-CoA transferase | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001193311.1 | Intron | NP_001180240.1 | ||||
NM_001193312.1 | Intron | NP_001180241.1 | ||||
NM_001193313.1 | Intron | NP_001180242.1 | ||||
NM_024728.2 | Intron | NP_079004.1 | ||||
XM_006715775.3 | Intron | XP_006715838.1 | ||||
XM_011515525.2 | Intron | XP_011513827.1 | ||||
XM_011515526.2 | Intron | XP_011513828.1 | ||||
XM_011515527.2 | Intron | XP_011513829.1 | ||||
XM_011515528.2 | Intron | XP_011513830.1 | ||||
XM_011515529.2 | Intron | XP_011513831.1 | ||||
XM_011515530.2 | Intron | XP_011513832.1 | ||||
XM_017012621.1 | Intron | XP_016868110.1 | ||||
XM_017012622.1 | Intron | XP_016868111.1 |
Set Membership: |
HapMap |