Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
TULP4 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
TULP4 - tubby like protein 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001007466.2 | Intron | NP_001007467.1 | ||||
NM_020245.4 | Intron | NP_064630.2 | ||||
XM_011535946.1 | Intron | XP_011534248.1 | ||||
XM_017011063.1 | Intron | XP_016866552.1 | ||||
XM_017011064.1 | Intron | XP_016866553.1 | ||||
XM_017011065.1 | Intron | XP_016866554.1 | ||||
XM_017011066.1 | Intron | XP_016866555.1 | ||||
XM_017011067.1 | Intron | XP_016866556.1 | ||||
XM_017011068.1 | Intron | XP_016866557.1 | ||||
XM_017011069.1 | Intron | XP_016866558.1 | ||||
XM_017011070.1 | Intron | XP_016866559.1 | ||||
XM_017011071.1 | Intron | XP_016866560.1 |
Set Membership: |
HapMap |