Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 604378 MIM: 614775 MIM: 601844 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
BECN1 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
BECN1 - beclin 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
CNTD1 - cyclin N-terminal domain containing 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_173478.2 | Intron | NP_775749.2 | ||||
XM_005257043.4 | Intron | XP_005257100.1 | ||||
XM_005257044.4 | Intron | XP_005257101.1 | ||||
XM_005257045.4 | Intron | XP_005257102.1 | ||||
XM_006721690.2 | Intron | XP_006721753.1 | ||||
XM_011524310.2 | Intron | XP_011522612.1 | ||||
XM_011524311.2 | Intron | XP_011522613.1 | ||||
XM_017024171.1 | Intron | XP_016879660.1 |
COA3 - cytochrome c oxidase assembly factor 3 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
WNK4 - WNK lysine deficient protein kinase 4 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |