Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCAGATGCAGCTGTCCAATCTTGAA[C/T]TTTCCAGCCATGAGAATCATGAACC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 108746 MIM: 609303 | ||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
ATP6V1E1 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
ATP6V1E1 - ATPase H+ transporting V1 subunit E1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001039366.1 | Intron | NP_001034455.1 | ||||
NM_001039367.1 | Intron | NP_001034456.1 | ||||
NM_001696.3 | Intron | NP_001687.1 |
LOC101929372 - uncharacterized LOC101929372 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SLC25A18 - solute carrier family 25 member 18 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |