Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTGGACTTTAGTGTCTGATCACCCT[C/T]TGAGGCTGGCATTGTCTCAAACCCA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 604800 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
DDX49 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
DDX49 - DEAD-box helicase 49 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
HOMER3 - homer scaffolding protein 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001145721.1 | Intron | NP_001139193.1 | ||||
NM_001145722.1 | Intron | NP_001139194.1 | ||||
NM_001145724.1 | Intron | NP_001139196.1 | ||||
NM_004838.3 | Intron | NP_004829.3 | ||||
XM_006722943.1 | Intron | XP_006723006.1 | ||||
XM_006722944.1 | Intron | XP_006723007.1 |
LOC102724360 - uncharacterized LOC102724360 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |