Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 138321 MIM: 607714 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
GPX3 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
GPX3 - glutathione peroxidase 3 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TNIP1 - TNFAIP3 interacting protein 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001252385.1 | 2302 | UTR 3 | NP_001239314.1 | |||
NM_001252386.1 | 2302 | UTR 3 | NP_001239315.1 | |||
NM_001252390.1 | 2302 | UTR 3 | NP_001239319.1 | |||
NM_001252391.1 | 2302 | UTR 3 | NP_001239320.1 | |||
NM_001252392.1 | 2302 | UTR 3 | NP_001239321.1 | |||
NM_001252393.1 | 2302 | UTR 3 | NP_001239322.1 | |||
NM_001258454.1 | 2302 | UTR 3 | NP_001245383.1 | |||
NM_001258455.1 | 2302 | UTR 3 | NP_001245384.1 | |||
NM_001258456.1 | 2302 | UTR 3 | NP_001245385.1 | |||
NM_006058.4 | 2302 | UTR 3 | NP_006049.3 | |||
XM_005268355.1 | 2302 | UTR 3 | XP_005268412.1 | |||
XM_006714751.1 | 2302 | UTR 3 | XP_006714814.1 | |||
XM_006714752.2 | 2302 | Intron | XP_006714815.1 | |||
XM_017008945.1 | 2302 | Intron | XP_016864434.1 | |||
XM_017008946.1 | 2302 | Intron | XP_016864435.1 | |||
XM_017008947.1 | 2302 | Intron | XP_016864436.1 | |||
XM_017008948.1 | 2302 | Intron | XP_016864437.1 | |||
XM_017008949.1 | 2302 | Intron | XP_016864438.1 | |||
XM_017008950.1 | 2302 | Intron | XP_016864439.1 | |||
XM_017008951.1 | 2302 | UTR 3 | XP_016864440.1 |
Set Membership: |
HapMap |