Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTATTTGTTTTCTCATGAGGAGAGT[A/G]TACTAGTAGTAGAAAAAAGTTACAT
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
CCDC144NL PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
CCDC144NL - coiled-coil domain containing 144 family, N-terminal like | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001004306.2 | Intron | NP_001004306.1 | ||||
XM_011523818.2 | Intron | XP_011522120.1 | ||||
XM_011523819.2 | Intron | XP_011522121.1 | ||||
XM_011523821.1 | Intron | XP_011522123.1 |
CCDC144NL-AS1 - CCDC144NL antisense RNA 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |