Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 611851 MIM: 611977 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
MRPL46 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
MRPL46 - mitochondrial ribosomal protein L46 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
MRPS11 - mitochondrial ribosomal protein S11 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001321970.1 | Intron | NP_001308899.1 | ||||
NM_001321972.1 | Intron | NP_001308901.1 | ||||
NM_001321973.1 | Intron | NP_001308902.1 | ||||
NM_001321974.1 | Intron | NP_001308903.1 | ||||
NM_001321976.1 | Intron | NP_001308905.1 | ||||
NM_022839.4 | Intron | NP_073750.2 | ||||
NM_176805.3 | Intron | NP_789775.1 |
Set Membership: |
HapMap |