Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
49 submissions
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 605176 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
HAO2 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
HAO2 - hydroxyacid oxidase 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001005783.2 | Intron | NP_001005783.2 | ||||
NM_016527.3 | Intron | NP_057611.1 | ||||
XM_011541558.1 | Intron | XP_011539860.1 | ||||
XM_011541559.1 | Intron | XP_011539861.1 | ||||
XM_011541560.1 | Intron | XP_011539862.1 | ||||
XM_011541561.1 | Intron | XP_011539863.1 | ||||
XM_017001421.1 | Intron | XP_016856910.1 |
HAO2-IT1 - HAO2 intronic transcript 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
HapMap |