Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACCTCAAACAAGGAAGACACCTCTT[A/G]GAGTTTCAGCTGTTACAACTCAGCT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 611395 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
FAM110C PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
FAM110C - family with sequence similarity 110 member C | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001077710.2 | 1431 | UTR 3 | NP_001071178.2 | |||
XM_011510372.2 | 1431 | UTR 3 | XP_011508674.1 | |||
XM_017004689.1 | 1431 | Intron | XP_016860178.1 | |||
XM_017004690.1 | 1431 | Intron | XP_016860179.1 | |||
XM_017004691.1 | 1431 | UTR 3 | XP_016860180.1 | |||
XM_017004692.1 | 1431 | Intron | XP_016860181.1 |
Set Membership: |
HapMap |