Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 147681 MIM: 603638 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
IL11 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
IL11 - interleukin 11 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
RPL28 - ribosomal protein L28 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TMEM190 - transmembrane protein 190 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_139172.1 | 173 | Intron | NP_631911.1 | |||
XM_017026331.1 | 173 | UTR 5 | XP_016881820.1 |
TMEM238 - transmembrane protein 238 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |