Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGAGGCTCATCAGGAGGTACTGTTG[C/G]AGTCTCAGCCTGAGGTGATGGGTGT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 605467 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
ZNF274 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
ZNF274 - zinc finger protein 274 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001278734.1 | Intron | NP_001265663.1 | ||||
NM_016324.3 | Intron | NP_057408.2 | ||||
NM_016325.3 | Intron | NP_057409.1 | ||||
NM_133502.2 | Intron | NP_598009.1 | ||||
XM_011526327.1 | Intron | XP_011524629.1 | ||||
XM_011526328.2 | Intron | XP_011524630.1 | ||||
XM_017026174.1 | Intron | XP_016881663.1 | ||||
XM_017026175.1 | Intron | XP_016881664.1 |
Set Membership: |
HapMap |