Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 605433 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
KIF13A PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
KIF13A - kinesin family member 13A | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001105566.2 | Intron | NP_001099036.1 | ||||
NM_001105567.2 | Intron | NP_001099037.1 | ||||
NM_001105568.2 | Intron | NP_001099038.1 | ||||
NM_001243423.1 | Intron | NP_001230352.1 | ||||
NM_022113.5 | Intron | NP_071396.4 | ||||
XM_011514806.2 | Intron | XP_011513108.1 | ||||
XM_017011188.1 | Intron | XP_016866677.1 | ||||
XM_017011189.1 | Intron | XP_016866678.1 | ||||
XM_017011190.1 | Intron | XP_016866679.1 | ||||
XM_017011191.1 | Intron | XP_016866680.1 | ||||
XM_017011192.1 | Intron | XP_016866681.1 | ||||
XM_017011193.1 | Intron | XP_016866682.1 |
Set Membership: |
HapMap |