Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
16 submissions
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
ZNF684 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
ZNF684 - zinc finger protein 684 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_152373.3 | 88 | Intron | NP_689586.3 | |||
XM_011540671.2 | 88 | UTR 5 | XP_011538973.1 | |||
XM_011540672.2 | 88 | UTR 5 | XP_011538974.1 | |||
XM_011540673.2 | 88 | Intron | XP_011538975.1 | |||
XM_017000290.1 | 88 | UTR 5 | XP_016855779.1 | |||
XM_017000291.1 | 88 | Intron | XP_016855780.1 | |||
XM_017000292.1 | 88 | UTR 5 | XP_016855781.1 |
Set Membership: |
HapMap |