Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
CCDC59 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CCDC59 - coiled-coil domain containing 59 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
METTL25 - methyltransferase like 25 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001319675.1 | Intron | NP_001306604.1 | ||||
NM_032230.2 | Intron | NP_115606.2 | ||||
XM_005269186.4 | Intron | XP_005269243.2 | ||||
XM_011538827.2 | Intron | XP_011537129.1 | ||||
XM_011538828.2 | Intron | XP_011537130.1 | ||||
XM_011538829.2 | Intron | XP_011537131.1 | ||||
XM_017020020.1 | Intron | XP_016875509.1 | ||||
XM_017020021.1 | Intron | XP_016875510.1 |