Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GTGTGGCTACTTCTATTAGAGCCCC[A/T]TGCTTCGCAGCTTCAGCCTTGCGGT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 172460 MIM: 194541 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
MTHFD1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
MTHFD1 - methylenetetrahydrofolate dehydrogenase, cyclohydrolase and formyltetrahydrofolate synthetase 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_005956.3 | Intron | NP_005947.3 |
ZBTB25 - zinc finger and BTB domain containing 25 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001304507.1 | Intron | NP_001291436.1 | ||||
NM_001304508.1 | Intron | NP_001291437.1 | ||||
NM_006977.3 | Intron | NP_008908.2 | ||||
XM_005268051.3 | Intron | XP_005268108.1 | ||||
XM_006720250.3 | Intron | XP_006720313.1 | ||||
XM_006720251.3 | Intron | XP_006720314.1 | ||||
XM_006720252.3 | Intron | XP_006720315.1 | ||||
XM_011537139.1 | Intron | XP_011535441.1 | ||||
XM_011537141.2 | Intron | XP_011535443.1 | ||||
XM_011537143.2 | Intron | XP_011535445.1 | ||||
XM_011537146.2 | Intron | XP_011535448.1 | ||||
XM_017021630.1 | Intron | XP_016877119.1 | ||||
XM_017021631.1 | Intron | XP_016877120.1 | ||||
XM_017021632.1 | Intron | XP_016877121.1 | ||||
XM_017021633.1 | Intron | XP_016877122.1 | ||||
XM_017021634.1 | Intron | XP_016877123.1 | ||||
XM_017021635.1 | Intron | XP_016877124.1 | ||||
XM_017021636.1 | Intron | XP_016877125.1 | ||||
XM_017021637.1 | Intron | XP_016877126.1 | ||||
XM_017021638.1 | Intron | XP_016877127.1 |