Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATTAGAGAGATAGTTGTGTAAGGAA[C/G]TTCTCATAATAAATTCTGAACTGTT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
ZNF394 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ZNF394 - zinc finger protein 394 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
ZNF789 - zinc finger protein 789 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001013258.1 | Intron | NP_001013276.1 | ||||
NM_213603.2 | Intron | NP_998768.2 | ||||
XM_011516067.2 | Intron | XP_011514369.1 | ||||
XM_011516070.2 | Intron | XP_011514372.1 | ||||
XM_011516073.2 | Intron | XP_011514375.1 | ||||
XM_017012018.1 | Intron | XP_016867507.1 | ||||
XM_017012019.1 | Intron | XP_016867508.1 | ||||
XM_017012020.1 | Intron | XP_016867509.1 | ||||
XM_017012021.1 | Intron | XP_016867510.1 | ||||
XM_017012022.1 | Intron | XP_016867511.1 | ||||
XM_017012023.1 | Intron | XP_016867512.1 | ||||
XM_017012024.1 | Intron | XP_016867513.1 | ||||
XM_017012025.1 | Intron | XP_016867514.1 | ||||
XM_017012026.1 | Intron | XP_016867515.1 | ||||
XM_017012027.1 | Intron | XP_016867516.1 |