Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCTAATCCATTCAGCACCTGGTGTG[A/G]TATTTGGTCTCACTTTGCCCTCCTG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
ZNF345 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
ZNF345 - zinc finger protein 345 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001242472.1 | Intron | NP_001229401.1 | ||||
NM_001242474.1 | Intron | NP_001229403.1 | ||||
NM_001242475.1 | Intron | NP_001229404.1 | ||||
NM_001242476.1 | Intron | NP_001229405.1 | ||||
NM_003419.4 | Intron | NP_003410.1 | ||||
XM_011526704.2 | Intron | XP_011525006.1 | ||||
XM_017026568.1 | Intron | XP_016882057.1 | ||||
XM_017026569.1 | Intron | XP_016882058.1 | ||||
XM_017026570.1 | Intron | XP_016882059.1 | ||||
XM_017026571.1 | Intron | XP_016882060.1 | ||||
XM_017026572.1 | Intron | XP_016882061.1 | ||||
XM_017026573.1 | Intron | XP_016882062.1 |
ZNF829 - zinc finger protein 829 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001037232.3 | Intron | NP_001032309.2 | ||||
NM_001171979.1 | Intron | NP_001165450.1 | ||||
XM_005258876.4 | Intron | XP_005258933.1 | ||||
XM_011526933.2 | Intron | XP_011525235.1 |
Set Membership: |
HapMap |