Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGATGCAAATTTCAGCCATTAAAAT[A/T]AAACTACTTAATAACTTAAATATTC
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 606698 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
SOX30 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
SOX30 - SRY-box 30 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001308165.1 | 1587 | UTR 3 | NP_001295094.1 | |||
NM_007017.2 | 1587 | UTR 3 | NP_008948.1 | |||
NM_178424.1 | 1587 | UTR 3 | NP_848511.1 | |||
XM_005265803.4 | 1587 | UTR 3 | XP_005265860.1 | |||
XM_011534420.1 | 1587 | UTR 3 | XP_011532722.1 |
Set Membership: |
HapMap |