Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGTACAAACTCAGGGCCCTAATTTT[C/T]TCAGCTGTAAAATGATCCGATCAGT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 607981 | ||||||||||||||||||||
Literature Links: |
NUB1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
NUB1 - negative regulator of ubiquitin like proteins 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001243351.1 | Intron | NP_001230280.1 | ||||
NM_016118.4 | Intron | NP_057202.3 | ||||
XM_005250011.2 | Intron | XP_005250068.1 | ||||
XM_017012304.1 | Intron | XP_016867793.1 | ||||
XM_017012305.1 | Intron | XP_016867794.1 | ||||
XM_017012306.1 | Intron | XP_016867795.1 | ||||
XM_017012307.1 | Intron | XP_016867796.1 | ||||
XM_017012308.1 | Intron | XP_016867797.1 | ||||
XM_017012309.1 | Intron | XP_016867798.1 | ||||
XM_017012310.1 | Intron | XP_016867799.1 | ||||
XM_017012311.1 | Intron | XP_016867800.1 | ||||
XM_017012312.1 | Intron | XP_016867801.1 | ||||
XM_017012313.1 | Intron | XP_016867802.1 | ||||
XM_017012314.1 | Intron | XP_016867803.1 |