Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 610335 | ||||||||||||||||||||
Literature Links: |
PHF20 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
PHF20 - PHD finger protein 20 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_016436.4 | Intron | NP_057520.2 | ||||
XM_005260421.2 | Intron | XP_005260478.1 | ||||
XM_011528843.1 | Intron | XP_011527145.1 | ||||
XM_011528844.1 | Intron | XP_011527146.1 | ||||
XM_017027864.1 | Intron | XP_016883353.1 | ||||
XM_017027865.1 | Intron | XP_016883354.1 | ||||
XM_017027866.1 | Intron | XP_016883355.1 | ||||
XM_017027867.1 | Intron | XP_016883356.1 | ||||
XM_017027868.1 | Intron | XP_016883357.1 | ||||
XM_017027869.1 | Intron | XP_016883358.1 | ||||
XM_017027870.1 | Intron | XP_016883359.1 | ||||
XM_017027871.1 | Intron | XP_016883360.1 |