Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGTTGTCAGAACACTGAGTGGATGG[C/T]GATGTCGCTGAAATGGAGAACGTGA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 609092 | ||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
FBXO10 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
FBXO10 - F-box protein 10 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_012166.2 | Intron | NP_036298.2 | ||||
XM_005251439.4 | Intron | XP_005251496.1 | ||||
XM_006716754.3 | Intron | XP_006716817.1 | ||||
XM_017014618.1 | Intron | XP_016870107.1 | ||||
XM_017014619.1 | Intron | XP_016870108.1 |
Set Membership: |
HapMap |