Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 616164 | ||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
SDR42E1 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
SDR42E1 - short chain dehydrogenase/reductase family 42E, member 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_145168.2 | 2048 | UTR 3 | NP_660151.2 | |||
XM_005256257.3 | 2048 | UTR 3 | XP_005256314.1 | |||
XM_011523471.2 | 2048 | UTR 3 | XP_011521773.1 | |||
XM_011523472.2 | 2048 | UTR 3 | XP_011521774.1 |
Set Membership: |
HapMap |