Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AAGCCTGCTCAGGTGGTCTCTTATA[C/T]GGATGCATGTAACATTACTGTAAAA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
ZNF846 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
ZNF846 - zinc finger protein 846 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001077624.1 | 2032 | Intron | NP_001071092.1 | |||
XM_005259772.3 | 2032 | Intron | XP_005259829.1 | |||
XM_005259773.4 | 2032 | Intron | XP_005259830.1 | |||
XM_005259775.3 | 2032 | Intron | XP_005259832.1 | |||
XM_006722658.3 | 2032 | UTR 3 | XP_006722721.1 | |||
XM_011527717.2 | 2032 | Intron | XP_011526019.1 | |||
XM_011527718.2 | 2032 | Intron | XP_011526020.1 | |||
XM_011527719.2 | 2032 | Intron | XP_011526021.1 | |||
XM_011527720.2 | 2032 | Intron | XP_011526022.1 | |||
XM_011527721.2 | 2032 | Intron | XP_011526023.1 | |||
XM_017026404.1 | 2032 | Intron | XP_016881893.1 | |||
XM_017026405.1 | 2032 | Intron | XP_016881894.1 | |||
XM_017026406.1 | 2032 | Intron | XP_016881895.1 | |||
XM_017026407.1 | 2032 | Intron | XP_016881896.1 | |||
XM_017026408.1 | 2032 | Intron | XP_016881897.1 |
Set Membership: |
HapMap |