Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 612408 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
PSPC1 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
PSPC1 - paraspeckle component 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001042414.2 | 2024 | Intron | NP_001035879.1 | |||
XM_006719844.3 | 2024 | Intron | XP_006719907.1 | |||
XM_011535137.1 | 2024 | Intron | XP_011533439.1 | |||
XM_011535138.2 | 2024 | UTR 3 | XP_011533440.1 | |||
XM_011535139.1 | 2024 | Intron | XP_011533441.1 | |||
XM_011535140.1 | 2024 | Intron | XP_011533442.1 | |||
XM_011535141.2 | 2024 | Intron | XP_011533443.1 | |||
XM_011535142.2 | 2024 | Intron | XP_011533444.1 | |||
XM_017020650.1 | 2024 | Intron | XP_016876139.1 |
Set Membership: |
HapMap |