Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGTGAGGCTTATGGGAGCTTCCATA[A/G]AAGTTGGGAGCAGGGACAAGACAGG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 601026 | ||||||||||||||||||||
Literature Links: |
AP2A1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
AP2A1 - adaptor related protein complex 2 alpha 1 subunit | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_014203.2 | 141 | Intron | NP_055018.2 | |||
NM_130787.2 | 141 | Intron | NP_570603.2 | |||
XM_011526556.2 | 141 | UTR 5 | XP_011524858.1 | |||
XM_011526557.2 | 141 | UTR 5 | XP_011524859.1 |