Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTTGGGACCAAGGTAGTCTGGCGCA[C/T]GGATGCCGAACCTTATCCGGGCTAG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 602919 MIM: 607163 | ||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
DOK1 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
DOK1 - docking protein 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001197260.1 | 669 | Intron | NP_001184189.1 | |||
NM_001318866.1 | 669 | Intron | NP_001305795.1 | |||
NM_001318867.1 | 669 | UTR 5 | NP_001305796.1 | |||
NM_001318868.1 | 669 | Intron | NP_001305797.1 | |||
NM_001318869.1 | 669 | Intron | NP_001305798.1 | |||
NM_001381.4 | 669 | Intron | NP_001372.1 | |||
XM_017003528.1 | 669 | Intron | XP_016859017.1 |
LOXL3 - lysyl oxidase like 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001289164.1 | 669 | Intron | NP_001276093.1 | |||
NM_001289165.1 | 669 | Intron | NP_001276094.1 | |||
NM_032603.3 | 669 | Intron | NP_115992.1 | |||
XM_011533134.2 | 669 | Intron | XP_011531436.1 | |||
XM_017005112.1 | 669 | Intron | XP_016860601.1 |
M1AP - meiosis 1 associated protein | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
HapMap |