Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 610061 | |||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
DNAH7 PubMed Links | |||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
DNAH7 - dynein axonemal heavy chain 7 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_018897.2 | Intron | NP_061720.2 | ||||
XM_011511487.2 | Intron | XP_011509789.1 | ||||
XM_011511488.2 | Intron | XP_011509790.1 | ||||
XM_011511489.2 | Intron | XP_011509791.1 | ||||
XM_011511490.2 | Intron | XP_011509792.1 | ||||
XM_011511491.2 | Intron | XP_011509793.1 | ||||
XM_011511492.2 | Intron | XP_011509794.1 | ||||
XM_011511493.2 | Intron | XP_011509795.1 | ||||
XM_011511494.2 | Intron | XP_011509796.1 | ||||
XM_011511495.2 | Intron | XP_011509797.1 | ||||
XM_011511497.2 | Intron | XP_011509799.1 | ||||
XM_017004504.1 | Intron | XP_016859993.1 |
Set Membership: |
HapMap |