Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTTTTGGAACATCACATGAGATGAG[A/G]GTATATAAAAGTGCCTAGCACAGTT
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 608185 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
EXOC4 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
EXOC4 - exocyst complex component 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001037126.1 | Intron | NP_001032203.1 | ||||
NM_021807.3 | Intron | NP_068579.3 | ||||
XM_005250523.4 | Intron | XP_005250580.1 | ||||
XM_017012494.1 | Intron | XP_016867983.1 | ||||
XM_017012495.1 | Intron | XP_016867984.1 |
Set Membership: |
HapMap |