Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GTTTAGCACAGTGAAGCATTTGCTA[C/T]GTGATGACATTTATTATGGTTAAAG
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 609584 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
ATP5S PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
ATP5S - ATP synthase, H+ transporting, mitochondrial Fo complex subunit s (factor B) | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001003803.2 | 376 | Intron | NP_001003803.1 | |||
NM_001003805.2 | 376 | Intron | NP_001003805.1 | |||
NM_015684.3 | 376 | Intron | NP_056499.2 | |||
XM_005267537.3 | 376 | Intron | XP_005267594.1 | |||
XM_011536649.2 | 376 | UTR 5 | XP_011534951.1 | |||
XM_011536650.2 | 376 | UTR 5 | XP_011534952.1 | |||
XM_011536651.2 | 376 | UTR 5 | XP_011534953.1 | |||
XM_011536652.1 | 376 | Intron | XP_011534954.1 | |||
XM_011536653.2 | 376 | Intron | XP_011534955.1 | |||
XM_011536654.2 | 376 | UTR 5 | XP_011534956.1 | |||
XM_011536655.2 | 376 | UTR 5 | XP_011534957.1 | |||
XM_011536656.2 | 376 | Intron | XP_011534958.1 | |||
XM_011536657.2 | 376 | Intron | XP_011534959.1 | |||
XM_017021220.1 | 376 | Intron | XP_016876709.1 |
L2HGDH - L-2-hydroxyglutarate dehydrogenase | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
HapMap |