Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 610512 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
SEC23B PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
SEC23B - Sec23 homolog B, coat complex II component | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001172745.1 | 535 | Missense Mutation | CCT,CTT | P,L 25 | NP_001166216.1 | |
NM_001172746.1 | 535 | Missense Mutation | CCT,CTT | P,L 25 | NP_001166217.1 | |
NM_006363.4 | 535 | Missense Mutation | CCT,CTT | P,L 25 | NP_006354.2 | |
NM_032985.4 | 535 | Missense Mutation | CCT,CTT | P,L 25 | NP_116780.1 | |
NM_032986.3 | 535 | Missense Mutation | CCT,CTT | P,L 25 | NP_116781.1 | |
XM_017027593.1 | 535 | Missense Mutation | CCT,CTT | P,L 25 | XP_016883082.1 |