Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GAAGATTCATCTGTAAGTGTTTACA[C/T]AGGCGTATGTTTTTATTTCATTTGG
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 613377 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
PLAC8L1 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
PLAC8L1 - PLAC8 like 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001029869.1 | Intron | NP_001025040.1 | ||||
XM_005268381.3 | Intron | XP_005268438.1 | ||||
XM_011537571.2 | Intron | XP_011535873.1 | ||||
XM_017009137.1 | Intron | XP_016864626.1 |
SH3RF2 - SH3 domain containing ring finger 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |