Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 601959 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
NEK4 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
NEK4 - NIMA related kinase 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001193533.1 | Intron | NP_001180462.1 | ||||
NM_003157.4 | Intron | NP_003148.2 | ||||
XM_006713310.2 | Intron | XP_006713373.1 | ||||
XM_011534039.2 | Intron | XP_011532341.1 | ||||
XM_011534040.2 | Intron | XP_011532342.1 | ||||
XM_017007085.1 | Intron | XP_016862574.1 | ||||
XM_017007086.1 | Intron | XP_016862575.1 |