Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 608669 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
BNC2 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
BNC2 - basonuclin 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001317939.1 | 16301 | UTR 3 | NP_001304868.1 | |||
NM_001317940.1 | 16301 | UTR 3 | NP_001304869.1 | |||
NM_017637.5 | 16301 | UTR 3 | NP_060107.3 | |||
XM_011517924.1 | 16301 | UTR 3 | XP_011516226.1 | |||
XM_011517934.2 | 16301 | Intron | XP_011516236.1 | |||
XM_017014816.1 | 16301 | UTR 3 | XP_016870305.1 | |||
XM_017014817.1 | 16301 | UTR 3 | XP_016870306.1 | |||
XM_017014818.1 | 16301 | UTR 3 | XP_016870307.1 | |||
XM_017014819.1 | 16301 | Intron | XP_016870308.1 | |||
XM_017014820.1 | 16301 | UTR 3 | XP_016870309.1 | |||
XM_017014821.1 | 16301 | Intron | XP_016870310.1 | |||
XM_017014822.1 | 16301 | UTR 3 | XP_016870311.1 | |||
XM_017014823.1 | 16301 | UTR 3 | XP_016870312.1 | |||
XM_017014824.1 | 16301 | UTR 3 | XP_016870313.1 | |||
XM_017014825.1 | 16301 | UTR 3 | XP_016870314.1 | |||
XM_017014826.1 | 16301 | Intron | XP_016870315.1 | |||
XM_017014827.1 | 16301 | Intron | XP_016870316.1 | |||
XM_017014828.1 | 16301 | Intron | XP_016870317.1 | |||
XM_017014829.1 | 16301 | UTR 3 | XP_016870318.1 |
Set Membership: |
HapMap |