Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
WDYHV1 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
WDYHV1 - WDYHV motif containing 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001283024.1 | Intron | NP_001269953.1 | ||||
NM_001283027.1 | Intron | NP_001269956.1 | ||||
NM_018024.2 | Intron | NP_060494.1 | ||||
XM_006716597.3 | Intron | XP_006716660.1 | ||||
XM_011517147.1 | Intron | XP_011515449.1 | ||||
XM_011517151.2 | Intron | XP_011515453.1 | ||||
XM_017013601.1 | Intron | XP_016869090.1 | ||||
XM_017013602.1 | Intron | XP_016869091.1 | ||||
XM_017013603.1 | Intron | XP_016869092.1 |