Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
5 submissions
|
||||||||||||||||||||
Phenotype: |
MIM: 602636 | ||||||||||||||||||||
Literature Links: |
PPP1R8 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
PPP1R8 - protein phosphatase 1 regulatory subunit 8 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_002713.3 | 182 | UTR 5 | NP_002704.1 | |||
NM_014110.4 | 182 | Intron | NP_054829.2 | |||
NM_138558.2 | 182 | UTR 5 | NP_612568.1 | |||
XM_017001594.1 | 182 | UTR 5 | XP_016857083.1 |
SCARNA1 - small Cajal body-specific RNA 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |