Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 607246 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
AP3D1 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
AP3D1 - adaptor related protein complex 3 delta 1 subunit | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001261826.1 | Intron | NP_001248755.1 | ||||
NM_003938.6 | Intron | NP_003929.4 | ||||
XM_006722932.2 | Intron | XP_006722995.1 | ||||
XM_017027422.1 | Intron | XP_016882911.1 |
IZUMO4 - IZUMO family member 4 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
MOB3A - MOB kinase activator 3A | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
HapMap |