Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 605147 MIM: 604098 | |||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
LECT1 PubMed Links | |||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese)
|
||||||
EUR
|
||||||||
AMR
|
LECT1 - leukocyte cell derived chemotaxin 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001011705.1 | 1084 | UTR 3 | NP_001011705.1 | |||
NM_007015.2 | 1084 | UTR 3 | NP_008946.1 | |||
XM_011534897.2 | 1084 | UTR 3 | XP_011533199.1 | |||
XM_011534898.2 | 1084 | UTR 3 | XP_011533200.1 | |||
XM_011534899.2 | 1084 | UTR 3 | XP_011533201.1 | |||
XM_011534900.2 | 1084 | UTR 3 | XP_011533202.1 |
SUGT1 - SGT1 homolog, MIS12 kinetochore complex assembly cochaperone | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |