Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTCGTATCTCCCTCTGCTAACCACT[G/T]TCTATTGTGACTTCTGTTGTGTGGT
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 605502 MIM: 154365 MIM: 604943 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
DNAJC2 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
DNAJC2 - DnaJ heat shock protein family (Hsp40) member C2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
PSMC2 - proteasome 26S subunit, ATPase 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001204453.1 | Intron | NP_001191382.1 | ||||
NM_002803.3 | Intron | NP_002794.1 | ||||
XM_005250505.1 | Intron | XP_005250562.1 |
SLC26A5 - solute carrier family 26 member 5 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001167962.1 | Intron | NP_001161434.1 | ||||
NM_001321787.1 | Intron | NP_001308716.1 | ||||
NM_198999.2 | Intron | NP_945350.1 | ||||
NM_206883.2 | Intron | NP_996766.1 | ||||
NM_206884.2 | Intron | NP_996767.1 | ||||
NM_206885.2 | Intron | NP_996768.1 | ||||
XM_011516170.2 | Intron | XP_011514472.1 |
Set Membership: |
HapMap |