Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 601688 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
HPGD PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
HPGD - hydroxyprostaglandin dehydrogenase 15-(NAD) | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000860.5 | 2132 | UTR 3 | NP_000851.2 | |||
NM_001145816.2 | 2132 | UTR 3 | NP_001139288.1 | |||
NM_001256301.1 | 2132 | UTR 3 | NP_001243230.1 | |||
NM_001256305.1 | 2132 | UTR 3 | NP_001243234.1 | |||
NM_001256306.1 | 2132 | UTR 3 | NP_001243235.1 | |||
NM_001256307.1 | 2132 | UTR 3 | NP_001243236.1 | |||
XM_011531907.1 | 2132 | Intron | XP_011530209.1 |
Set Membership: |
HapMap |