Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GTCACTACAGATGATTCCTTTATGC[C/T]AACCCTGCTGGGAAAATATTTATTG
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
CLEC4F PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
CLEC4F - C-type lectin domain family 4 member F | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001258027.1 | Intron | NP_001244956.1 | ||||
NM_001321308.1 | Intron | NP_001308237.1 | ||||
NM_173535.2 | Intron | NP_775806.2 | ||||
XM_011532635.2 | Intron | XP_011530937.1 | ||||
XM_011532637.2 | Intron | XP_011530939.1 | ||||
XM_011532638.2 | Intron | XP_011530940.1 | ||||
XM_011532639.2 | Intron | XP_011530941.1 | ||||
XM_011532640.2 | Intron | XP_011530942.1 | ||||
XM_011532641.2 | Intron | XP_011530943.1 | ||||
XM_011532642.2 | Intron | XP_011530944.1 | ||||
XM_011532643.1 | Intron | XP_011530945.1 | ||||
XM_017003519.1 | Intron | XP_016859008.1 |
Set Membership: |
HapMap |