Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATATTGCAGAACCCTACAACAGCCC[C/T]CAAGTGGGAAGAGTAAGATTGAGAG
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
LONRF1 PubMed Links | |||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese)
|
||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
LONRF1 - LON peptidase N-terminal domain and ring finger 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
XM_005273685.3 | Intron | XP_005273742.1 | ||||
XM_011544694.2 | Intron | XP_011542996.1 |
MIR3926-1 - microRNA 3926-1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
MIR3926-2 - microRNA 3926-2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
MIR5692A2 - microRNA 5692a-2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
HapMap |