Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCGGGGGGCAGGGTCCCGGGCGGGC[A/G]CGGGCGGCTCGGCAGGCTTGCTCAA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
30 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
C1orf159 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
C1orf159 - chromosome 1 open reading frame 159 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
LOC100288175 - uncharacterized LOC100288175 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
LOC105378948 - uncharacterized LOC105378948 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
RNF223 - ring finger protein 223 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001205252.1 | 816 | Missense Mutation | GCG,GTG | A,V 172 | NP_001192181.1 | |
XM_017001286.1 | 816 | Missense Mutation | GCG,GTG | A,V 172 | XP_016856775.1 | |
XM_017001287.1 | 816 | Intron | XP_016856776.1 |