Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TAATTTACCAAATTCCATATTTAGA[C/T]TGGGGTGGGGAGAGAGGCTCTCTCG
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 614797 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
PELI1 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
PELI1 - pellino E3 ubiquitin protein ligase 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_020651.3 | 2947 | UTR 3 | NP_065702.2 | |||
XM_005264437.3 | 2947 | UTR 3 | XP_005264494.1 | |||
XM_011532994.2 | 2947 | UTR 3 | XP_011531296.1 | |||
XM_011532996.2 | 2947 | UTR 3 | XP_011531298.1 | |||
XM_011532997.2 | 2947 | UTR 3 | XP_011531299.1 | |||
XM_017004520.1 | 2947 | UTR 3 | XP_016860009.1 | |||
XM_017004521.1 | 2947 | UTR 3 | XP_016860010.1 |
Set Membership: |
HapMap |