Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 604960 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
PACSIN2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
PACSIN2 - protein kinase C and casein kinase substrate in neurons 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001184970.1 | 2333 | UTR 3 | NP_001171899.1 | |||
NM_001184971.1 | 2333 | UTR 3 | NP_001171900.1 | |||
NM_007229.3 | 2333 | UTR 3 | NP_009160.2 | |||
XM_005261319.3 | 2333 | UTR 3 | XP_005261376.1 | |||
XM_011529846.1 | 2333 | UTR 3 | XP_011528148.1 | |||
XM_011529847.2 | 2333 | UTR 3 | XP_011528149.1 | |||
XM_011529848.2 | 2333 | UTR 3 | XP_011528150.1 | |||
XM_011529849.2 | 2333 | UTR 3 | XP_011528151.1 | |||
XM_011529850.2 | 2333 | UTR 3 | XP_011528152.1 | |||
XM_011529851.2 | 2333 | UTR 3 | XP_011528153.1 | |||
XM_011529852.1 | 2333 | UTR 3 | XP_011528154.1 | |||
XM_011529853.1 | 2333 | UTR 3 | XP_011528155.1 | |||
XM_017028562.1 | 2333 | UTR 3 | XP_016884051.1 | |||
XM_017028563.1 | 2333 | UTR 3 | XP_016884052.1 | |||
XM_017028564.1 | 2333 | UTR 3 | XP_016884053.1 | |||
XM_017028565.1 | 2333 | UTR 3 | XP_016884054.1 | |||
XM_017028566.1 | 2333 | UTR 3 | XP_016884055.1 | |||
XM_017028567.1 | 2333 | UTR 3 | XP_016884056.1 | |||
XM_017028568.1 | 2333 | UTR 3 | XP_016884057.1 |