Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 600405 | |||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
RFC3 PubMed Links | |||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
RFC3 - replication factor C subunit 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_002915.3 | Intron | NP_002906.1 | ||||
NM_181558.2 | Intron | NP_853536.2 | ||||
XM_011535172.2 | Intron | XP_011533474.1 | ||||
XM_011535173.2 | Intron | XP_011533475.1 | ||||
XM_011535174.2 | Intron | XP_011533476.1 | ||||
XM_011535175.1 | Intron | XP_011533477.1 | ||||
XM_017020680.1 | Intron | XP_016876169.1 | ||||
XM_017020681.1 | Intron | XP_016876170.1 | ||||
XM_017020682.1 | Intron | XP_016876171.1 | ||||
XM_017020683.1 | Intron | XP_016876172.1 |
Set Membership: |
HapMap |