Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGCTTTGTTGATGTCCCTGGAAGCA[A/G]CTGCCCATATCATTATAAATTATAT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 607043 | ||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
TRAF3IP2 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
TRAF3IP2 - TRAF3 interacting protein 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001164281.2 | 2727 | UTR 3 | NP_001157753.1 | |||
NM_001164283.2 | 2727 | UTR 3 | NP_001157755.1 | |||
NM_147200.2 | 2727 | Intron | NP_671733.2 | |||
NM_147686.3 | 2727 | UTR 3 | NP_679211.2 | |||
XM_006715319.3 | 2727 | Intron | XP_006715382.1 | |||
XM_011535386.1 | 2727 | Intron | XP_011533688.1 |
TRAF3IP2-AS1 - TRAF3IP2 antisense RNA 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
HapMap |