Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 615614 | ||||||||||||||||||||
Literature Links: |
KLHL32 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
MMS22L - MMS22 like, DNA repair protein | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_198468.2 | 7173 | UTR 3 | NP_940870.2 | |||
XM_006715430.1 | 7173 | UTR 3 | XP_006715493.1 | |||
XM_006715432.3 | 7173 | Intron | XP_006715495.1 | |||
XM_011535670.2 | 7173 | Intron | XP_011533972.1 | |||
XM_011535671.2 | 7173 | Intron | XP_011533973.1 | |||
XM_011535672.2 | 7173 | Intron | XP_011533974.1 | |||
XM_011535673.2 | 7173 | Intron | XP_011533975.1 | |||
XM_011535674.2 | 7173 | Intron | XP_011533976.1 | |||
XM_011535675.2 | 7173 | Intron | XP_011533977.1 | |||
XM_011535676.2 | 7173 | Intron | XP_011533978.1 | |||
XM_011535677.2 | 7173 | Intron | XP_011533979.1 | |||
XM_011535678.2 | 7173 | Intron | XP_011533980.1 | |||
XM_011535679.2 | 7173 | Intron | XP_011533981.1 | |||
XM_011535680.2 | 7173 | Intron | XP_011533982.1 | |||
XM_017010656.1 | 7173 | Intron | XP_016866145.1 | |||
XM_017010657.1 | 7173 | Intron | XP_016866146.1 |