Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGGGCGCCAGGCACATGGTGAGGGA[C/T]ACCAGGTAGCCATTAGAAACGGCAA
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 602110 | |||||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
LOC102724064 PubMed Links | |||||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU) - Not Available | |||||||||
EAS
|
African American
|
YRI (Yoruba)
|
|||||||||
SAS
|
Japanese
|
CHB (Han Chinese) - Not Available | |||||||||
AFR
|
Chinese
|
JPT (Japanese) - Not Available | |||||||||
EUR
|
|||||||||||
AMR
|
LOC102724064 - uncharacterized LOC102724064 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SLC29A2 - solute carrier family 29 member 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001300868.1 | 1613 | Silent Mutation | GTA,GTG | V,V 411 | NP_001287797.1 | |
NM_001300869.1 | 1613 | UTR 3 | NP_001287798.1 | |||
NM_001532.2 | 1613 | Silent Mutation | GTA,GTG | V,V 411 | NP_001523.2 | |
XM_011544974.1 | 1613 | UTR 3 | XP_011543276.1 | |||
XM_017017630.1 | 1613 | UTR 3 | XP_016873119.1 | |||
XM_017017631.1 | 1613 | Silent Mutation | GTA,GTG | V,V 411 | XP_016873120.1 | |
XM_017017632.1 | 1613 | Silent Mutation | GTA,GTG | V,V 411 | XP_016873121.1 | |
XM_017017633.1 | 1613 | Intron | XP_016873122.1 | |||
XM_017017634.1 | 1613 | Silent Mutation | GTA,GTG | V,V 353 | XP_016873123.1 | |
XM_017017635.1 | 1613 | Silent Mutation | GTA,GTG | V,V 353 | XP_016873124.1 | |
XM_017017636.1 | 1613 | Intron | XP_016873125.1 | |||
XM_017017637.1 | 1613 | UTR 3 | XP_016873126.1 | |||
XM_017017638.1 | 1613 | UTR 3 | XP_016873127.1 |
Set Membership: |
HapMap DME Validated Inventoried |